Assay Details
Target Gene Details
Entrez Gene ID: | 84168 |
Gene Name: | anthrax toxin receptor 1 |
Gene Aliases: |
ATR, GAPO, TEM8 |
Location: |
Chr.2:69013144-69249327 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANTXR1 | NM_018153.3 | NP_060623.2 | ||
| NM_032208.2 | NP_115584.1 | |||
| NM_053034.2 | NP_444262.1 | |||
| XM_017005075.1 | XP_016860564.1 | |||
| XM_017005076.1 | XP_016860565.1 | |||
| XM_017005077.1 | XP_016860566.1 | |||
| AF279145.2 | AAK52094.1 | |||
| AF421380.1 | AAL26496.1 | |||
| AK001463.1 | BAA91707.1 | |||
| AK223273.1 | BAD96993.1 | |||
| AK292113.1 | ||||
| AK309553.1 | ||||
| BC012074.1 | AAH12074.1 | |||
| CF126791.1 | ||||
| DC360415.1 | ||||
| DC361072.1 | ||||
| JX424838.1 | ||||
| JX424839.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs113599325] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map