Assay Details
Target Gene Details
Entrez Gene ID: | 9759 |
Gene Name: | histone deacetylase 4 |
Gene Aliases: |
AHO3, BDMR, HA6116, HD4, HDAC-4, HDAC-A, HDACA |
Location: |
Chr.2:239048168-239401647 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HDAC4 | NM_006037.3 | NP_006028.2 | ||
| XM_006712878.3 | XP_006712941.1 | |||
| XM_011512221.1 | XP_011510523.1 | |||
| XM_011512222.2 | XP_011510524.1 | |||
| XM_011512223.2 | XP_011510525.1 | |||
| XM_011512226.2 | XP_011510528.1 | |||
| AB006626.2 | ||||
| AF132607.1 | AAD29046.1 | |||
| AK308791.1 | ||||
| BC039904.1 | AAH39904.1 |
Target Gene Details
Entrez Gene ID: | 100616360 |
Gene Name: | microRNA 2467 |
Gene Aliases: |
mir-2467 |
Location: |
Chr.2:239351724-239351804 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR2467 | NR_039948.1 |
More Information
Set Membership: |
|

Genomic Map