Assay Details
Target Gene Details
Entrez Gene ID: | 100506866 |
Gene Name: | TTN antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.2:178522827-178620217 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTN-AS1 | NR_038271.1 | |||
| NR_038272.1 | ||||
| AK093733.1 | ||||
| DB229458.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834468 | Chr.2:178486389 - 178668898 on Build GRCh38 | Gain |
|
| nsv834467 | Chr.2:178393413 - 178566238 on Build GRCh38 | Loss |
|
| nsv954437 | Chr.2:178523774 - 178531073 on Build GRCh38 | Duplication |
|

Genomic Map