Assay Details
Target Gene Details
Entrez Gene ID: | 64895 |
Gene Name: | poly(A) polymerase gamma |
Gene Aliases: |
- |
Location: |
Chr.2:60756230-60802086 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAPOLG | NM_022894.3 | NP_075045.2 | ||
| XM_005264500.3 | XP_005264557.1 | |||
| XM_005264501.1 | XP_005264558.1 | |||
| XM_011533041.1 | XP_011531343.1 | |||
| XM_017004723.1 | XP_016860212.1 | |||
| XM_017004724.1 | XP_016860213.1 | |||
| AF312211.1 | AAK83701.1 | |||
| AJ308101.1 | CAC59751.1 | |||
| AK021867.1 | BAB13919.1 | |||
| AK314663.1 | ||||
| AY029162.1 | AAK31791.1 | |||
| BC016738.1 | ||||
| BC110906.1 | AAI10907.1 | |||
| BC111701.1 | AAI11702.1 | |||
| BC113747.1 | AAI13748.1 | |||
| BX647824.1 | ||||
| DB082179.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3859n100 | Chr.2:60754968 - 60868884 on Build GRCh38 | Gain |
|
| dgv6908n54 | Chr.2:60756834 - 60892336 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76296742] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map