Assay Details
Target Gene Details
Entrez Gene ID: | 5746 |
Gene Name: | parathyroid hormone 2 receptor |
Gene Aliases: |
PTHR2 |
Location: |
Chr.2:208359845-208494506 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTH2R | NM_001309516.1 | NP_001296445.1 | ||
| AK294181.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv584297 | Chr.2:208155457 - 208452035 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map