Assay Details
Target Gene Details
Entrez Gene ID: | 79901 |
Gene Name: | cytochrome b reductase 1 |
Gene Aliases: |
CYB561A2, DCYTB, FRRS3 |
Location: |
Chr.2:171522247-171558133 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYBRD1 | NM_001127383.1 | NP_001120855.1 | ||
| NM_001256909.1 | NP_001243838.1 | |||
| NM_024843.3 | NP_079119.3 | |||
| AK027115.1 | BAB15661.1 | |||
| AK128495.1 | BAC87466.1 | |||
| AK225459.1 | ||||
| AK291731.1 | ||||
| AK301485.1 | ||||
| AK316588.1 | ||||
| AL136693.1 | CAB66628.1 | |||
| BC065290.1 | AAH65290.1 | |||
| CB266594.1 | ||||
| CX866825.1 | ||||
| DB235339.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160884 | Chr.2:171536097 - 171553747 on Build GRCh38 | Duplication |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78424806] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map