Assay Details
Target Gene Details
Entrez Gene ID: | 6670 |
Gene Name: | Sp3 transcription factor |
Gene Aliases: |
SPR2 |
Location: |
Chr.2:173906459-173965702 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SP3 | NM_001017371.4 | NP_001017371.3 | ||
| NM_001172712.1 | NP_001166183.1 | |||
| NM_003111.4 | NP_003102.1 | |||
| AB209334.1 | BAD92571.1 | |||
| AK304199.1 | ||||
| AY070137.1 | AAL58086.1 | |||
| AY441957.1 | AAR30505.1 | |||
| AY441958.1 | AAR30506.1 | |||
| BC042945.2 | AAH42945.2 | |||
| BC126414.1 | ||||
| BC143928.1 | ||||
| M97191.2 | AAA36630.2 | |||
| X68560.1 | CAA48562.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv751e214 | Chr.2:173920627 - 173932730 on Build GRCh38 | Loss |
|
| nsv953518 | Chr.2:173922873 - 173963172 on Build GRCh38 | Duplication |
|
| esv3593394 | Chr.2:173921104 - 173932730 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73028236,rs73028237] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map