Assay Details
Target Gene Details
Entrez Gene ID: | 8520 |
Gene Name: | histone acetyltransferase 1 |
Gene Aliases: |
KAT1 |
Location: |
Chr.2:171922425-171983686 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HAT1 | NM_003642.3 | NP_003633.1 | ||
| NR_027862.1 | ||||
| XM_006712808.3 | XP_006712871.1 | |||
| XM_011512027.2 | 1 | 678 | XP_011510329.1 | |
| XM_017005136.1 | XP_016860625.1 | |||
| XM_017005137.1 | XP_016860626.1 | |||
| AF030424.1 | AAC02425.1 | |||
| AK309001.1 | ||||
| AK313699.1 | ||||
| BC063003.1 | AAH63003.1 | |||
| DB050336.1 |
More Information
Set Membership: |
|

Genomic Map