Assay Details
Target Gene Details
Entrez Gene ID: | 33 |
Gene Name: | acyl-CoA dehydrogenase, long chain |
Gene Aliases: |
ACAD4, LCAD |
Location: |
Chr.2:210187990-210225491 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACADL | NM_001608.3 | NP_001599.1 | ||
| XM_005246517.4 | XP_005246574.1 | |||
| XM_017003955.1 | XP_016859444.1 | |||
| AK313498.1 | ||||
| BC039063.1 | AAH39063.1 | |||
| BC064549.1 | AAH64549.1 | |||
| M74096.1 | AAA51565.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs79293046] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map