Assay Details
Target Gene Details
Entrez Gene ID: | 9173 |
Gene Name: | interleukin 1 receptor like 1 |
Gene Aliases: |
DER4, FIT-1, IL33R, ST2, ST2L, ST2V, T1 |
Location: |
Chr.2:102311502-102352367 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL1RL1 | NM_001282408.1 | NP_001269337.1 | ||
| NM_016232.4 | NP_057316.3 | |||
| XM_011512151.1 | XP_011510453.1 | |||
| AB012701.3 | ||||
| AK303389.1 |
More Information
Set Membership: |
|

Genomic Map