Assay Details
Target Gene Details
Entrez Gene ID: | 391356 |
Gene Name: | peptidyl-tRNA hydrolase domain containing 1 |
Gene Aliases: |
C2orf79 |
Location: |
Chr.2:24790267-24793382 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTRHD1 | NM_001013663.1 | NP_001013685.1 | ||
| BC073803.1 | AAH73803.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1004529 | Chr.2:24363242 - 25005468 on Build GRCh38 | Loss |
|
| nsv508831 | Chr.2:24787088 - 24857561 on Build GRCh38 | Insertion |
|
| nsv997725 | Chr.2:24052436 - 25123406 on Build GRCh38 | Gain |
|
| nsv1000020 | Chr.2:24709475 - 24909903 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map