Assay Details
Target Gene Details
Entrez Gene ID: | 130502 |
Gene Name: | tetratricopeptide repeat domain 32 |
Gene Aliases: |
- |
Location: |
Chr.2:19896631-19901986 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC32 | NM_001008237.2 | NP_001008238.1 | ||
| BC057850.1 | AAH57850.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521619 | Chr.2:19898447 - 19951056 on Build GRCh38 | Loss |
|
| esv29979 | Chr.2:19408886 - 20481173 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map