Assay Details
Target Gene Details
Entrez Gene ID: | 51455 |
Gene Name: | REV1, DNA directed polymerase |
Gene Aliases: |
AIBP80, REV1L |
Location: |
Chr.2:99400475-99490123 on Build GRCh38 |
Assay Gene Location: | Within Intron 25 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524466 | Chr.2:99335623 - 99415089 on Build GRCh38 | Loss |
|
| dgv4032n100 | Chr.2:99364388 - 99481422 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map