Assay Details
Target Gene Details
Entrez Gene ID: | 57162 |
Gene Name: | pellino E3 ubiquitin protein ligase 1 |
Gene Aliases: |
- |
Location: |
Chr.2:64092652-64144471 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PELI1 | NM_020651.3 | NP_065702.2 | ||
| XM_005264437.3 | XP_005264494.1 | |||
| XM_011532994.2 | XP_011531296.1 | |||
| XM_011532996.2 | XP_011531298.1 | |||
| XM_011532997.2 | XP_011531299.1 | |||
| XM_017004520.1 | XP_016860009.1 | |||
| XM_017004521.1 | XP_016860010.1 | |||
| AF300987.1 | AAG17451.1 | |||
| AF302505.1 | AAG15393.1 | |||
| AJ278859.1 | CAC04320.1 | |||
| AK312441.1 | ||||
| AY364257.1 | AAQ76816.1 | |||
| BC050019.1 | AAH50019.1 | |||
| BC063611.1 | AAH63611.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv151e55 | Chr.2:64085008 - 64294232 on Build GRCh38 | Gain |
|
| nsv582127 | Chr.2:63618350 - 64923736 on Build GRCh38 | Gain |
|
| nsv1013132 | Chr.2:64008041 - 64403950 on Build GRCh38 | Loss |
|
| nsv834243 | Chr.2:64089380 - 64249801 on Build GRCh38 | Gain+Loss |
|
| esv2759055 | Chr.2:64025871 - 64312422 on Build GRCh38 | Gain |
|
| nsv582126 | Chr.2:63346276 - 64470570 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114303054] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map