Assay Details
Target Gene Details
Entrez Gene ID: | 285154 |
Gene Name: | CYP1B1 antisense RNA 1 |
Gene Aliases: |
C2orf58 |
Location: |
Chr.2:38131105-38181855 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP1B1-AS1 | NR_027252.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv581476 | Chr.2:38086798 - 38160085 on Build GRCh38 | Loss |
|
| nsv581477 | Chr.2:38117986 - 38150263 on Build GRCh38 | Loss |
|
| nsv9646 | Chr.2:38131169 - 38855047 on Build GRCh38 | Loss |
|

Genomic Map