Assay Details
Target Gene Details
Entrez Gene ID: | 10130 |
Gene Name: | protein disulfide isomerase family A member 6 |
Gene Aliases: |
ERP5, P5, TXNDC7 |
Location: |
Chr.2:10783391-10837977 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDIA6 | NM_001282704.1 | NP_001269633.1 | ||
| NM_001282705.1 | NP_001269634.1 | |||
| NM_001282706.1 | NP_001269635.1 | |||
| NM_001282707.1 | NP_001269636.1 | |||
| NM_005742.3 | NP_005733.1 | |||
| XM_011510308.1 | XP_011508610.1 | |||
| AK125449.1 | 1 | 429 | ||
| AK127433.1 | BAC86977.1 | |||
| AK131234.1 | ||||
| AK225496.1 | ||||
| AK289428.1 | ||||
| AK294347.1 | ||||
| AK297547.1 | ||||
| BC001312.1 | AAH01312.1 | |||
| U79278.1 | AAB50217.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1000731 | Chr.2:10618531 - 10803756 on Build GRCh38 | Gain |
|
| nsv580968 | Chr.2:10783142 - 10793123 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73170277] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map