Assay Details
Target Gene Details
Entrez Gene ID: | 114805 |
Gene Name: | polypeptide N-acetylgalactosaminyltransferase 13 |
Gene Aliases: |
GalNAc-T13 |
Location: |
Chr.2:153871898-154454177 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GALNT13 | NM_052917.3 | NP_443149.2 | ||
| XM_011510537.2 | XP_011508839.1 | |||
| XM_017003258.1 | XP_016858747.1 | |||
| XM_017003260.1 | XP_016858749.1 | |||
| XM_017003261.1 | XP_016858750.1 | |||
| AK131195.1 | ||||
| CN267103.1 | ||||
| HY107944.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv583367 | Chr.2:153834664 - 153892470 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72862073] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map