Assay Details
Target Gene Details
Entrez Gene ID: | 23671 |
Gene Name: | transmembrane protein with EGF like and two follistatin like domains 2 |
Gene Aliases: |
CT120.2, HPP1, TENB2, TPEF, TR, TR-2 |
Location: |
Chr.2:191948299-192194940 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEFF2 | NM_001305134.1 | NP_001292063.1 | ||
| NM_016192.3 | NP_057276.2 | |||
| XM_017003739.1 | 9 | 2799 | XP_016859228.1 | |
| AL157430.1 | CAB75654.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv584068 | Chr.2:191838120 - 192003002 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map