Assay Details
Target Gene Details
Entrez Gene ID: | 79828 |
Gene Name: | methyltransferase like 8 |
Gene Aliases: |
TIP |
Location: |
Chr.2:171315746-171434803 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| METTL8 | NM_001321154.1 | 10 | 8491 | NP_001308083.1 |
| NM_001321155.1 | 11 | 8615 | NP_001308084.1 | |
| NM_001321156.1 | 11 | 8685 | NP_001308085.1 | |
| NM_001321157.1 | 11 | 8518 | NP_001308086.1 | |
| NM_001321158.1 | 9 | 8239 | NP_001308087.1 | |
| NM_001321159.1 | 9 | 8336 | NP_001308088.1 | |
| NM_001321160.1 | 10 | 8508 | NP_001308089.1 | |
| NM_001321161.1 | 8 | 8132 | NP_001308090.1 | |
| NM_001321162.1 | 9 | 8353 | NP_001308091.1 | |
| NM_024770.4 | 10 | 8394 | NP_079046.2 | |
| NR_135568.1 | 7 | 7736 |
More Information
Set Membership: |
|

Genomic Map