Assay Details
Target Gene Details
Entrez Gene ID: | 79083 |
Gene Name: | melanophilin |
Gene Aliases: |
SLAC2-A |
Location: |
Chr.2:237486410-237555318 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MLPH | NM_001281474.1 | NP_001268403.1 | ||
| AK094168.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv584728 | Chr.2:237362083 - 237525791 on Build GRCh38 | Loss |
|
| nsv1000252 | Chr.2:235899999 - 238766417 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map