Assay Details
Target Gene Details
Entrez Gene ID: | 3938 |
Gene Name: | lactase |
Gene Aliases: |
LAC, LPH, LPH1 |
Location: |
Chr.2:135787840-135837200 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LCT | NM_002299.3 | NP_002290.2 | ||
| XM_017004088.1 | XP_016859577.1 | |||
| X07994.1 | CAA30801.1 |
Target Gene Details
Entrez Gene ID: | 100507600 |
Gene Name: | uncharacterized LOC100507600 |
Gene Aliases: |
- |
Location: |
Chr.2:135820191-135823087 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100507600 | NR_045486.1 | 1 | 362 | |
| AK097922.1 | 1 | 364 |
More Information
Set Membership: |
|

Genomic Map