Assay Details
Target Gene Details
Entrez Gene ID: | 200576 |
Gene Name: | phosphoinositide kinase, FYVE-type zinc finger containing |
Gene Aliases: |
CFD, FAB1, HEL37, PIP5K, PIP5K3, ZFYVE29 |
Location: |
Chr.2:208266178-208358751 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv584297 | Chr.2:208155457 - 208452035 on Build GRCh38 | Gain |
|
| nsv834517 | Chr.2:208142073 - 208305545 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map