Assay Details
Target Gene Details
Entrez Gene ID: | 57007 |
Gene Name: | atypical chemokine receptor 3 |
Gene Aliases: |
CMKOR1, CXC-R7, CXCR-7, CXCR7, GPR159, RDC-1, RDC1 |
Location: |
Chr.2:236537131-236582358 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACKR3 | XM_005246098.3 | XP_005246155.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2576763 | Chr.2:236542403 - 236543488 on Build GRCh38 | Insertion |
|
| nsv1005677 | Chr.2:236369512 - 236811991 on Build GRCh38 | Gain |
|
| nsv997438 | Chr.2:236210757 - 237034862 on Build GRCh38 | Gain |
|
| nsv1000252 | Chr.2:235899999 - 238766417 on Build GRCh38 | Loss |
|
| nsv834576 | Chr.2:236466567 - 236654210 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map