Assay Details
Target Gene Details
Entrez Gene ID: | 7447 |
Gene Name: | visinin like 1 |
Gene Aliases: |
HLP3, HPCAL3, HUVISL1, VILIP, VILIP-1 |
Location: |
Chr.2:17540540-17656439 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
VSNL1 | NM_003385.4 | NP_003376.2 | ||
AB001104.1 | BAA86891.1 | |||
AF039555.1 | AAD02174.1 | |||
BC022012.2 | AAH22012.1 | |||
BI544338.1 | ||||
U14747.1 | AAA91295.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv997502 | Chr.2:17160707 - 18147073 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
