Assay Details
Target Gene Details
Entrez Gene ID: | 7341 |
Gene Name: | small ubiquitin-like modifier 1 |
Gene Aliases: |
DAP1, GMP1, OFC10, PIC1, SENP2, SMT3, SMT3C, SMT3H3, UBL1 |
Location: |
Chr.2:202206180-202238599 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SUMO1 | NM_001005781.1 | NP_001005781.1 | ||
| NM_001005782.1 | NP_001005782.1 | |||
| NM_003352.4 | NP_003343.1 | |||
| AB062294.1 | BAB93477.1 | |||
| AK311840.1 | ||||
| BC006462.1 | AAH06462.1 | |||
| BC053528.1 | AAH53528.1 | |||
| BC066306.1 | AAH66306.1 | |||
| BG503431.1 | ||||
| BG569821.1 | ||||
| U38784.1 | AAC50733.1 | |||
| U61397.1 | AAB40388.1 | |||
| U67122.1 | AAC50996.1 | |||
| U83117.1 | AAB39999.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834512 | Chr.2:202100718 - 202263238 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map