Assay Details
Target Gene Details
Entrez Gene ID: | 25797 |
Gene Name: | glutaminyl-peptide cyclotransferase |
Gene Aliases: |
GCT, QC, sQC |
Location: |
Chr.2:37344610-37373322 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
QPCT | NM_012413.3 | NP_036545.1 | ||
AK290605.1 | ||||
BC036721.1 | AAH36721.1 | |||
BC047756.2 | AAH47756.1 | |||
BI755122.1 | ||||
X71125.1 | CAA50438.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv2686 | Chr.2:37347524 - 37380543 on Build GRCh38 | Insertion |
![]() |
nsv2685 | Chr.2:37322953 - 37367780 on Build GRCh38 | Deletion |
![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
