Assay Details
Target Gene Details
Entrez Gene ID: | 9173 |
Gene Name: | interleukin 1 receptor like 1 |
Gene Aliases: |
DER4, FIT-1, IL33R, ST2, ST2L, ST2V, T1 |
Location: |
Chr.2:102311502-102352367 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL1RL1 | NM_001282408.1 | NP_001269337.1 | ||
| NM_016232.4 | NP_057316.3 | |||
| XM_011512151.1 | XP_011510453.1 | |||
| AB012701.3 | ||||
| AK303389.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2673990 | Chr.2:102314830 - 102320561 on Build GRCh38 | Deletion |
|
| esv3591787 | Chr.2:102314919 - 102320486 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78496732] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map