Assay Details
Target Gene Details
Entrez Gene ID: | 23178 |
Gene Name: | PAS domain containing serine/threonine kinase |
Gene Aliases: |
PASKIN, STK37 |
Location: |
Chr.2:241106099-241150347 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575345 | Chr.2:241086207 - 241153068 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map