Assay Details
Target Gene Details
Entrez Gene ID: | 117581 |
Gene Name: | twist family bHLH transcription factor 2 |
Gene Aliases: |
AMS, BBRSAY, DERMO1, FFDD3, SETLSS, bHLHa39 |
Location: |
Chr.2:238848032-238910548 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TWIST2 | NM_001271893.3 | NP_001258822.1 | ||
| NM_057179.2 | NP_476527.1 | |||
| AW072674.1 | ||||
| BC017907.1 | AAH17907.1 | |||
| BC033168.1 | AAH33168.1 | |||
| BC103755.1 | AAI03756.1 | |||
| BU615717.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3594773 | Chr.2:238869973 - 238892363 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map