Assay Details
Target Gene Details
Entrez Gene ID: | 57520 |
Gene Name: | HECT, C2 and WW domain containing E3 ubiquitin protein ligase 2 |
Gene Aliases: |
NEDL2 |
Location: |
Chr.2:196194382-196593692 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HECW2 | NM_001304840.1 | NP_001291769.1 | ||
| NM_020760.2 | NP_065811.1 | |||
| XM_006712646.3 | XP_006712709.1 | |||
| XM_006712648.3 | XP_006712711.1 | |||
| XM_017004563.1 | XP_016860052.1 | |||
| AB037722.1 | ||||
| BC117194.1 | ||||
| BC117198.1 | ||||
| CR749424.1 | CAH18262.1 |
Target Gene Details
Entrez Gene ID: | 101927482 |
Gene Name: | uncharacterized LOC101927482 |
Gene Aliases: |
- |
Location: |
Chr.2:196260024-196264204 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927482 | NR_110225.1 | |||
| NR_110226.1 | ||||
| AK023739.1 | ||||
| CD638300.1 | ||||
| CD641185.1 |
More Information
Set Membership: |
|

Genomic Map