Assay Details
Target Gene Details
Entrez Gene ID: | 4118 |
Gene Name: | mal, T-cell differentiation protein |
Gene Aliases: |
- |
Location: |
Chr.2:95025655-95053992 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAL | NM_002371.3 | NP_002362.1 | ||
| NM_022438.2 | NP_071883.1 | |||
| NM_022439.2 | NP_071884.1 | |||
| NM_022440.2 | NP_071885.1 | |||
| AK311844.1 | ||||
| BC000458.2 | AAH00458.1 | |||
| BC003006.1 | AAH03006.1 | |||
| CR541879.1 | CAG46677.1 | |||
| DT215613.1 | ||||
| M15800.1 | AAA36196.1 | |||
| X76678.1 | CAA54100.1 | |||
| X76679.1 | CAA54101.1 | |||
| X76680.1 | CAA54102.1 | |||
| X76681.1 | CAA54103.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3583922 | Chr.2:95046007 - 95069566 on Build GRCh38 | Loss |
|
| dgv4007n100 | Chr.2:94662150 - 95248719 on Build GRCh38 | Gain |
|
| esv3591662 | Chr.2:94986856 - 95146757 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map