Assay Details
Target Gene Details
Entrez Gene ID: | 8291 |
Gene Name: | dysferlin |
Gene Aliases: |
FER1L1, LGMD2B, MMD1 |
Location: |
Chr.2:71453155-71686763 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DYSF | NM_001130976.1 | NP_001124448.1 | ||
| NM_001130977.1 | NP_001124449.1 | |||
| NM_001130978.1 | NP_001124450.1 | |||
| NM_001130979.1 | NP_001124451.1 | |||
| NM_001130980.1 | NP_001124452.1 | |||
| NM_001130981.1 | NP_001124453.1 | |||
| NM_003494.3 | NP_003485.1 | |||
| XM_005264585.4 | XP_005264642.1 | |||
| AF075575.1 | AAC63519.1 | |||
| EU515155.1 | ||||
| EU515156.1 | ||||
| EU515157.1 | ||||
| EU515158.1 | ||||
| EU515159.1 | ||||
| EU515160.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3591230 | Chr.2:71457775 - 71458754 on Build GRCh38 | Loss |
|
| nsv521345 | Chr.2:71345325 - 71458399 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map