Assay Details
Target Gene Details
Entrez Gene ID: | 3694 |
Gene Name: | integrin subunit beta 6 |
Gene Aliases: |
AI1H |
Location: |
Chr.2:160099666-160200313 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITGB6 | NM_000888.4 | NP_000879.2 | ||
| NM_001282353.1 | NP_001269282.1 | |||
| NM_001282354.1 | NP_001269283.1 | |||
| NM_001282355.1 | NP_001269284.1 | |||
| NM_001282388.1 | NP_001269317.1 | |||
| NM_001282389.1 | NP_001269318.1 | |||
| NM_001282390.1 | NP_001269319.1 | |||
| AK290300.1 | ||||
| AK304203.1 | ||||
| AK313944.1 | ||||
| AL831998.1 | ||||
| BC121178.2 | ||||
| CD013944.1 | ||||
| CD013945.1 | ||||
| CD013946.1 | ||||
| CD013947.1 | ||||
| M35198.3 | AAA36122.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1010774 | Chr.2:159012247 - 160175703 on Build GRCh38 | Gain |
|
| nsv583497 | Chr.2:160107092 - 160222924 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map