Assay Details
Target Gene Details
Entrez Gene ID: | 8291 |
Gene Name: | dysferlin |
Gene Aliases: |
FER1L1, LGMD2B, MMD1 |
Location: |
Chr.2:71453155-71686763 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Entrez Gene ID: | 8291 |
Gene Name: | dysferlin |
Gene Aliases: |
FER1L1, LGMD2B, MMD1 |
Location: |
Chr.2:71453155-71686763 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Set Membership: |
|