Assay Details
Target Gene Details
Entrez Gene ID: | 9781 |
Gene Name: | ring finger protein 144A |
Gene Aliases: |
RNF144, UBCE7IP4 |
Location: |
Chr.2:6917392-7076886 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF144A | NM_014746.4 | NP_055561.2 | ||
| XM_005246200.3 | XP_005246257.1 | |||
| XM_017005396.1 | XP_016860885.1 | |||
| XM_017005397.1 | XP_016860886.1 | |||
| XM_017005398.1 | XP_016860887.1 | |||
| XM_017005399.1 | XP_016860888.1 | |||
| XM_017005400.1 | XP_016860889.1 | |||
| XM_017005401.1 | XP_016860890.1 | |||
| XM_017005402.1 | XP_016860891.1 | |||
| XM_017005403.1 | XP_016860892.1 | |||
| XM_017005404.1 | XP_016860893.1 | |||
| AK293854.1 | ||||
| BC050373.1 | AAH50373.1 | |||
| CD101678.1 | ||||
| D79983.1 | ||||
| DA234807.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833326 | Chr.2:6844896 - 7002433 on Build GRCh38 | Loss |
|
| nsv2597 | Chr.2:6929286 - 6960834 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map