Assay Details
Target Gene Details
Entrez Gene ID: | 285154 |
Gene Name: | CYP1B1 antisense RNA 1 |
Gene Aliases: |
C2orf58 |
Location: |
Chr.2:38131105-38181855 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP1B1-AS1 | NR_027252.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv6788n54 | Chr.2:38148887 - 38177244 on Build GRCh38 | Loss |
|
| nsv9646 | Chr.2:38131169 - 38855047 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs66760537] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map