Assay Details
Target Gene Details
Entrez Gene ID: | 114805 |
Gene Name: | polypeptide N-acetylgalactosaminyltransferase 13 |
Gene Aliases: |
GalNAc-T13 |
Location: |
Chr.2:153871898-154454177 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GALNT13 | NM_001301627.1 | NP_001288556.1 | ||
| NM_052917.3 | NP_443149.2 | |||
| XM_011510537.2 | XP_011508839.1 | |||
| XM_011510538.2 | XP_011508840.1 | |||
| XM_017003258.1 | XP_016858747.1 | |||
| XM_017003259.1 | XP_016858748.1 | |||
| XM_017003260.1 | XP_016858749.1 | |||
| XM_017003261.1 | XP_016858750.1 | |||
| XM_017003262.1 | XP_016858751.1 | |||
| XM_017003263.1 | XP_016858752.1 | |||
| AB067505.1 | BAB67811.1 | |||
| AB078142.1 | BAC54545.1 | |||
| AJ505991.2 | CAD44533.2 | |||
| AK131195.1 | ||||
| AK308615.1 | ||||
| BC101031.1 | AAI01032.1 | |||
| BC101032.2 | ||||
| BC101033.1 | AAI01034.1 | |||
| BC101034.2 | ||||
| BG221433.1 | ||||
| KJ534843.1 | ||||
| KJ535058.1 |
Target Gene Details
Entrez Gene ID: | 100144595 |
Gene Name: | uncharacterized LOC100144595 |
Gene Aliases: |
- |
Location: |
Chr.2:154435853-154457438 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100144595 | NM_001256368.1 | NP_001243297.1 | ||
| AK090913.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv583421 | Chr.2:154401008 - 154524026 on Build GRCh38 | Loss |
|
| nsv583422 | Chr.2:154406489 - 154475974 on Build GRCh38 | Loss |
|
| esv3592991 | Chr.2:154445647 - 154453382 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map