Assay Details
Target Gene Details
Entrez Gene ID: | 64080 |
Gene Name: | ribokinase |
Gene Aliases: |
RBSK, RK |
Location: |
Chr.2:27781364-27890396 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBKS | NM_001287580.1 | NP_001274509.1 | ||
| NM_022128.2 | NP_071411.1 | |||
| AJ404857.1 | CAC12877.1 | |||
| AY643715.1 | ||||
| BC017425.1 | AAH17425.1 | |||
| BX648909.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs79290457] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map