Assay Details
Target Gene Details
Entrez Gene ID: | 103689911 |
Gene Name: | CATIP antisense RNA 2 |
Gene Aliases: |
- |
Location: |
Chr.2:218326241-218357966 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CATIP-AS2 | NR_125777.1 | |||
| CD672221.1 |
Target Gene Details
Entrez Gene ID: | 25953 |
Gene Name: | paroxysmal nonkinesigenic dyskinesia |
Gene Aliases: |
BRP17, DYT8, FKSG19, FPD1, KIPP1184, MR-1, MR1, PDC, PKND1, TAHCCP2 |
Location: |
Chr.2:218270392-218346793 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PNKD | NM_015488.4 | NP_056303.3 | ||
| NM_022572.4 | NP_072094.1 | |||
| XM_017003771.1 | XP_016859260.1 | |||
| XM_017003772.1 | XP_016859261.1 | |||
| AB033010.1 | BAA86498.1 | |||
| AK289867.1 | ||||
| AY358753.1 | AAQ89113.1 | |||
| BC002937.2 | AAH02937.1 | |||
| BC036457.2 | AAH36457.1 | |||
| BI764334.1 | ||||
| BU553179.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519829 | Chr.2:218286495 - 218403058 on Build GRCh38 | Loss |
|
| esv3594336 | Chr.2:218316567 - 218347668 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map