Assay Details
Target Gene Details
Entrez Gene ID: | 101926982 |
Gene Name: | protein GVQW2-like |
Gene Aliases: |
- |
Location: |
Chr.2:9649308-9665708 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101926982 | XM_005246205.1 | XP_005246262.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1011802 | Chr.2:9626042 - 9668155 on Build GRCh38 | Gain |
|
| nsv580915 | Chr.2:9168019 - 9704049 on Build GRCh38 | Gain |
|
| esv3589778 | Chr.2:9649326 - 9672251 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73913160] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map