Assay Details
Target Gene Details
Entrez Gene ID: | 101928103 |
Gene Name: | uncharacterized LOC101928103 |
Gene Aliases: |
- |
Location: |
Chr.2:214810229-214963605 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928103 | NR_110292.1 | |||
| AK056813.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1008103 | Chr.2:214811481 - 214858268 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs376147657] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map