Assay Details
Target Gene Details
Entrez Gene ID: | 64342 |
Gene Name: | HCLS1 binding protein 3 |
Gene Aliases: |
ETM2, HS1-BP3 |
Location: |
Chr.2:20553861-20651661 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HS1BP3 | XM_017004696.1 | XP_016860185.1 | ||
| XM_017004697.1 | XP_016860186.1 | |||
| XM_017004698.1 | XP_016860187.1 | |||
| XM_017004699.1 | XP_016860188.1 |
Target Gene Details
Entrez Gene ID: | 100874343 |
Gene Name: | HS1BP3 intronic transcript 1 |
Gene Aliases: |
- |
Location: |
Chr.2:20590775-20592548 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HS1BP3-IT1 | NR_046836.1 | 2 | 220 | |
| BQ010717.1 | 1 | 397 | ||
| BQ013798.1 | 1 | 397 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv56n68 | Chr.2:20430331 - 20664318 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map