Assay Details
Target Gene Details
Entrez Gene ID: | 5937 |
Gene Name: | RNA binding motif single stranded interacting protein 1 |
Gene Aliases: |
C2orf12, HCC-4, MSSP, MSSP-1, MSSP-2, MSSP-3, SCR2, YC1 |
Location: |
Chr.2:160272151-160493856 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBMS1 | NM_002897.4 | 14 | 2181 | NP_002888.1 |
| NM_016836.3 | 14 | 2190 | NP_058520.1 | |
| XM_005246737.1 | 15 | 1949 | XP_005246794.1 | |
| XM_005246738.2 | 15 | 2284 | XP_005246795.1 | |
| XM_005246739.2 | 15 | 2154 | XP_005246796.1 | |
| XM_005246740.1 | 15 | 1940 | XP_005246797.1 | |
| XM_005246741.3 | 15 | 2151 | XP_005246798.1 | |
| XM_017004626.1 | 14 | 2227 | XP_016860115.1 | |
| BC018951.2 | 14 | 2177 | AAH18951.1 | |
| CN399839.1 |
More Information
Set Membership: |
|

Genomic Map