Assay Details
Target Gene Details
Entrez Gene ID: | 9125 |
Gene Name: | CCR4-NOT transcription complex subunit 9 |
Gene Aliases: |
CAF40, CT129, RCD-1, RCD1, RQCD1 |
Location: |
Chr.2:218568579-218596435 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNOT9 | NM_001271634.1 | NP_001258563.1 | ||
| NM_001271635.1 | NP_001258564.1 | |||
| NM_005444.2 | NP_005435.1 | |||
| NR_073390.1 | ||||
| XM_011512138.2 | XP_011510440.1 | |||
| XM_017005249.1 | XP_016860738.1 | |||
| AB500892.1 | ||||
| AK293281.1 | ||||
| AK316567.1 | ||||
| BC007102.1 | ||||
| BC137455.1 | ||||
| BC137456.1 | ||||
| DC299874.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3168 | Chr.2:218536749 - 218582278 on Build GRCh38 | Deletion |
|
| nsv1131549 | Chr.2:218557668 - 218573059 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map