Assay Details
Target Gene Details
Entrez Gene ID: | 5062 |
Gene Name: | p21 (RAC1) activated kinase 2 |
Gene Aliases: |
PAK65, PAKgamma |
Location: |
Chr.3:196739857-196832647 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAK2 | NM_002577.4 | NP_002568.2 | ||
| XM_017006501.1 | XP_016861990.1 | |||
| AI140979.1 | ||||
| AK291339.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1009975 | Chr.3:196680410 - 196822871 on Build GRCh38 | Gain |
|
| nsv1001045 | Chr.3:196703082 - 196900384 on Build GRCh38 | Gain |
|
| nsv1003135 | Chr.3:196621850 - 197012082 on Build GRCh38 | Gain |
|
| nsv1014692 | Chr.3:196515378 - 196804953 on Build GRCh38 | Gain |
|
| esv2763775 | Chr.3:196727069 - 197015419 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map