Assay Details
Target Gene Details
Entrez Gene ID: | 55779 |
Gene Name: | cilia and flagella associated protein 44 |
Gene Aliases: |
WDR52 |
Location: |
Chr.3:113286930-113445190 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CFAP44 | NM_001164496.1 | NP_001157968.1 | ||
| NM_018338.3 | NP_060808.2 | |||
| XM_006713696.2 | XP_006713759.1 | |||
| XM_011512977.2 | XP_011511279.1 | |||
| XM_011512979.2 | XP_011511281.1 | |||
| XM_011512980.2 | XP_011511282.1 | |||
| XM_011512981.1 | XP_011511283.1 | |||
| XM_011512982.2 | XP_011511284.1 | |||
| XM_011512983.2 | XP_011511285.1 | |||
| XM_011512984.2 | XP_011511286.1 | |||
| XM_017006842.1 | XP_016862331.1 | |||
| XM_017006843.1 | XP_016862332.1 | |||
| XM_017006844.1 | XP_016862333.1 | |||
| XM_017006845.1 | XP_016862334.1 | |||
| XM_017006848.1 | XP_016862337.1 | |||
| XM_017006849.1 | XP_016862338.1 | |||
| AK056472.1 | BAB71193.1 | |||
| BC101582.1 | AAI01583.1 | |||
| BC113553.1 | ||||
| DB050626.1 |
Target Gene Details
Entrez Gene ID: | 100874029 |
Gene Name: | CFAP44 antisense RNA 1 |
Gene Aliases: |
WDR52-AS1 |
Location: |
Chr.3:113403998-113433992 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CFAP44-AS1 | NR_046728.1 | |||
| BX108680.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1013506 | Chr.3:113171761 - 113962104 on Build GRCh38 | Gain |
|

Genomic Map