Assay Details
Target Gene Details
Entrez Gene ID: | 50650 |
Gene Name: | Rho guanine nucleotide exchange factor 3 |
Gene Aliases: |
GEF3, STA3, XPLN |
Location: |
Chr.3:56727418-57079308 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
More Information
Set Membership: |
|

Genomic Map