Assay Details
Target Gene Details
Entrez Gene ID: | 57337 |
Gene Name: | SUMO1/sentrin specific peptidase 7 |
Gene Aliases: |
- |
Location: |
Chr.3:101324189-101513241 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1003475 | Chr.3:101286394 - 101401186 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map