Assay Details
Target Gene Details
Entrez Gene ID: | 152404 |
Gene Name: | immunoglobulin superfamily member 11 |
Gene Aliases: |
BT-IgSF, CT119, CXADRL1, Igsf13, VSIG3 |
Location: |
Chr.3:118900630-119146068 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Target Gene Details
Entrez Gene ID: | 100506765 |
Gene Name: | IGSF11 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:118943076-118948241 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IGSF11-AS1 | NR_046230.1 | |||
| BI829854.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1011135 | Chr.3:116380194 - 119036472 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map