Assay Details
Target Gene Details
Entrez Gene ID: | 51161 |
Gene Name: | chromosome 3 open reading frame 18 |
Gene Aliases: |
G20 |
Location: |
Chr.3:50558025-50574835 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3584502 | Chr.3:50327055 - 50622310 on Build GRCh38 | Gain |
|
| nsv460533 | Chr.3:50536782 - 50640720 on Build GRCh38 | Loss |
|
| nsv590272 | Chr.3:50498842 - 50578618 on Build GRCh38 | Loss |
|

Genomic Map